Gene Transcripts
- Gene Symbol
- Thoc7
- Entrez Gene ID
- 66231
- Gene Name
- THO complex 7 homolog (Drosophila)
- Gene Aliases
- Nif3l1bp1
- Chromosome Location
- Chr. 14: 13949012 - 13962042 on Build GRCm38
- UniGene ID
- Mm.295875
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001285780.1 |
NP_001272709.1
|
3, 4 |
328 |
|
NM_025435.3 |
NP_079711.1
|
2, 3 |
250 |
|
GenBank mRNA |
AA409027.1 |
- |
2, 3 |
251 |
|
AK005176.1 |
- |
2, 3 |
177 |
|
AK007440.1 |
- |
2, 3 |
156 |
|
AK008899.1 |
- |
3, 4 |
233 |
|
AK010958.1 |
- |
2, 3 |
191 |
|
AK013319.1 |
- |
2, 3 |
173 |
|
AK147003.1 |
- |
3, 4 |
231 |
|
BC042209.1 |
- |
2, 3 |
177 |
|
BC054419.1 |
- |
2, 3 |
210 |
4010343 |
BC116919.1 |
- |
2, 3 |
215 |
40126238 |
BC116945.1 |
- |
2, 3 |
215 |
40126264 |
CK329337.1 |
- |
2, 3 |
251 |
|
View Full Product Details