Gene Transcripts
- Gene Symbol
- AGA
- Entrez Gene ID
- 175
- Gene Name
- aspartylglucosaminidase
- Gene Aliases
- AGU, ASRG, GA
- Chromosome Location
- Chr. 4: 177430774 - 177442503 on Build GRCh38
- UniGene ID
- Hs.207776
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000027.3 |
NP_000018.2
|
4, 5 |
642 |
|
NM_001171988.1 |
NP_001165459.1
|
4, 5 |
642 |
|
NR_033655.1 |
|
4, 5 |
642 |
|
GenBank mRNA |
AK303370.1 |
- |
3, 4 |
435 |
|
AK312982.1 |
- |
4, 5 |
576 |
|
BC012392.1 |
- |
4, 5 |
565 |
3859317 |
CX869892.1 |
- |
4, 5 |
519 |
|
M64073.1 |
- |
4, 5 |
517 |
|
M64075.1 |
- |
2, 3 |
210 |
|
M64076.1 |
- |
2, 3 |
210 |
|
X55762.1 |
- |
4, 5 |
541 |
|
View Full Product Details