Gene Transcripts

Gene Symbol
Rras2
Entrez Gene ID
66922
Gene Name
related RAS viral (r-ras) oncogene homolog 2
Gene Aliases
2610016H24Rik, C86394, TC21
Chromosome Location
Chr. 7: 114046782 - 114117781 on Build GRCm38
UniGene ID
Mm.276572
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025846.2 NP_080122.2 3 505
GenBank mRNA AK011419.1 - 3 505
AK078870.1 - 3 506
AK172037.1 - 3 497
AK172113.1 - 3 491
AK216768.1 - 1 18
BC003871.1 - 3 456 3492159

View Full Product Details