Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026360.3 | NP_080636.2 | 2, 3 | 219 | |
GenBank mRNA | AK010310.1 | - | 2, 3 | 212 | |
AK076982.1 | - | 2, 3 | 204 | ||
BC060168.1 | - | 2, 3 | 199 | ||
BC095944.1 | - | 2, 3 | 196 | 4935970 | |
BC132242.1 | - | 2, 3 | 211 | 40130519 | |
BC132244.1 | - | 2, 3 | 211 | 40130521 | |
BY123864.1 | - | 2, 3 | 219 |