Gene Transcripts
- Gene Symbol
- Nop9
- Entrez Gene ID
- 67842
- Gene Name
- NOP9 nucleolar protein
- Gene Aliases
- 2610027L16Rik
- Chromosome Location
- Chr. 14: 55745693 - 55755634 on Build GRCm38
- UniGene ID
- Mm.180338
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026403.3 |
NP_080679.3
|
3, 4 |
887 |
|
GenBank mRNA |
AK011570.1 |
- |
3, 4 |
877 |
|
AK015585.1 |
- |
3, 4 |
637 |
|
AK032873.1 |
- |
3, 4 |
871 |
|
AK037831.1 |
- |
3, 4 |
875 |
|
AK050912.1 |
- |
3, 4 |
878 |
|
AK083861.1 |
- |
3, 4 |
756 |
|
AK139689.1 |
- |
3, 4 |
838 |
|
AK146020.1 |
- |
3, 4 |
842 |
|
AK146687.1 |
- |
3, 4 |
859 |
|
AK152732.1 |
- |
3, 4 |
861 |
|
AK154387.1 |
- |
3, 4 |
879 |
|
AK159798.1 |
- |
3, 4 |
861 |
|
BC018523.1 |
- |
3, 4 |
867 |
3499986 |
View Full Product Details