Gene Transcripts
- Gene Symbol
- Ccdc34
- Entrez Gene ID
- 68201
- Gene Name
- coiled-coil domain containing 34
- Gene Aliases
- 2810027O19Rik, 4930522P10Rik, AI596202
- Chromosome Location
- Chr. 2: 110017510 - 110173355 on Build GRCm38
- UniGene ID
- Mm.181767
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_026613.4 |
NP_080889.3
|
5, 6 |
948 |
|
GenBank mRNA |
AK011768.1 |
- |
4, 5 |
464 |
|
AK033555.1 |
- |
5, 6 |
927 |
|
AK140947.1 |
- |
2, 3 |
2856 |
|
AK147056.1 |
- |
5, 6 |
949 |
|
BC049229.1 |
- |
5, 6 |
681 |
|
BC089458.1 |
- |
7, 8 |
1113 |
|
BC108339.1 |
- |
5, 6 |
932 |
|
BC119347.1 |
- |
4, 5 |
493 |
8734096 |
BC120580.1 |
- |
4, 5 |
493 |
40129503 |
View Full Product Details