Gene Transcripts

Gene Symbol
Snx6
Entrez Gene ID
72183
Gene Name
sorting nexin 6
Gene Aliases
2010006G21Rik, 2610032J07Rik, 2810425K19Rik, AU018928, C85963
Chromosome Location
Chr. 12: 54746353 - 54795749 on Build GRCm38
UniGene ID
Mm.28240
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026998.3 NP_081274.2 2 145
GenBank mRNA AK011640.1 - 3 147
AK013158.1 - 2 146
AK182704.1 - 1 84
BC025911.1 - 2 132

View Full Product Details