Gene Transcripts
- Gene Symbol
- Haghl
- Entrez Gene ID
- 68977
- Gene Name
- hydroxyacylglutathione hydrolase-like
- Gene Aliases
- 1500017E18Rik, 2810014I23Rik, C330022E15Rik
- Chromosome Location
- Chr. 17: 25779843 - 25785669 on Build GRCm38
- UniGene ID
- Mm.29230
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001271433.1 |
NP_001258362.1
|
6, 7 |
757 |
|
NM_001271434.1 |
NP_001258363.1
|
6, 7 |
757 |
|
NM_001271435.1 |
NP_001258364.1
|
6, 7 |
757 |
|
NM_001271436.1 |
NP_001258365.1
|
5, 6 |
571 |
|
NM_001271437.1 |
NP_001258366.1
|
5, 6 |
566 |
|
NM_026897.3 |
NP_081173.1
|
5, 6 |
742 |
|
GenBank mRNA |
AK005274.1 |
- |
5, 6 |
738 |
|
AK012748.1 |
- |
6, 7 |
754 |
|
AK032999.1 |
- |
3, 4 |
715 |
|
AK034798.1 |
- |
6, 7 |
732 |
|
AK047517.1 |
- |
6, 7 |
712 |
|
AK079632.1 |
- |
3, 4 |
906 |
|
AK080454.1 |
- |
1 |
1693 |
|
AK154248.1 |
- |
5, 6 |
547 |
|
BC005586.1 |
- |
6, 7 |
703 |
|
BC030466.1 |
- |
6, 7 |
742 |
5253213 |
BC083322.1 |
- |
6, 7 |
696 |
6820885 |
View Full Product Details