Gene Transcripts
- Gene Symbol
- Spsb3
- Entrez Gene ID
- 79043
- Gene Name
- splA/ryanodine receptor domain and SOCS box containing 3
- Gene Aliases
- 2310012N15Rik, 3300001M01Rik, SSB3, Tce1, mKIAA4204
- Chromosome Location
- Chr. 17: 24886349 - 24892147 on Build GRCm38
- UniGene ID
- Mm.274794
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001163750.1 |
NP_001157222.1
|
3, 4 |
749 |
|
NM_001163751.1 |
NP_001157223.1
|
3, 4 |
404 |
|
NM_027141.2 |
NP_081417.2
|
3, 4 |
425 |
|
GenBank mRNA |
AF403038.1 |
- |
3, 4 |
425 |
|
AK009325.1 |
- |
3, 4 |
425 |
|
AK076170.1 |
- |
3, 4 |
701 |
|
AK082768.1 |
- |
3, 4 |
348 |
|
AK167474.1 |
- |
3, 4 |
665 |
|
AK184627.1 |
- |
1, 2 |
23 |
|
AK220234.1 |
- |
3, 4 |
687 |
|
BC072556.1 |
- |
3, 4 |
710 |
5691114 |
View Full Product Details