Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027288.3 | NP_081564.3 | 6 | 896 | |
GenBank mRNA | AF306557.1 | - | 6 | 900 | |
AK036505.1 | - | 7 | 996 | ||
AK047057.1 | - | 6 | 889 | ||
AK142165.1 | - | 6 | 861 | ||
AK154172.1 | - | 6 | 877 | ||
AK166022.1 | - | 6 | 878 | ||
AK166340.1 | - | 6 | 894 | ||
AK170290.1 | - | 6 | 893 | ||
BC031409.1 | - | 6 | 839 | 4952710 |