Gene Transcripts

Gene Symbol
Mier2
Entrez Gene ID
70427
Gene Name
mesoderm induction early response 1, family member 2
Gene Aliases
2700087H15Rik, mKIAA1193
Chromosome Location
Chr. 10: 79540245 - 79555631 on Build GRCm38
UniGene ID
Mm.334193
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027422.2 NP_081698.2 14 2505
GenBank mRNA AK012576.1 - 2 1353
AK154672.1 - 14 2522
AK173128.1 - 13 2660
AK178663.1 - 1 41
AK179152.1 - 1 41
BC005748.1 - 1 913
BC067013.1 - 14 2505 6810181
BC080757.1 - 7 1756
BC083321.1 - 5 1626

View Full Product Details