Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_028117.3 | NP_082393.3 | 1 | 350 | |
GenBank mRNA | AK011230.1 | - | 1 | 287 | |
AK154555.1 | - | 1 | 339 | ||
AK154636.1 | - | 1 | 319 | ||
AK155115.1 | - | 1 | 333 | ||
AK159357.1 | - | 1 | 318 | ||
AK171614.1 | - | 1 | 220 | ||
BC026886.1 | - | 1 | 171 | ||
BC043700.1 | - | 1 | 323 | 5346204 | |
BC085479.1 | - | 1 | 267 | 6415106 |