Gene Transcripts

Gene Symbol
Mon1a
Entrez Gene ID
72825
Gene Name
MON1 homolog A (yeast)
Gene Aliases
2810468K17Rik
Chromosome Location
Chr. 9: 107888112 - 107903139 on Build GRCm38
UniGene ID
Mm.38037
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028369.3 NP_082645.1 3 791
GenBank mRNA AK013387.1 - 3 738
AK172624.1 - 3 698
BC048852.1 - 3 717 5148425
BC058717.1 - 3 769 5704178

View Full Product Details