Gene Transcripts

Gene Symbol
Speer4b
Entrez Gene ID
73526
Gene Name
spermatogenesis associated glutamate (E)-rich protein 4b
Gene Aliases
1700081O22Rik, SPEER-4B
Chromosome Location
Chr. 5: 27495809 - 27501392 on Build GRCm38
UniGene ID
Mm.389608
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_028561.2 NP_082837.1 4 543
GenBank mRNA AK006975.1 - 4 592
BC119075.1 - 4 543 8733824

View Full Product Details