Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001102436.1 | NP_001095906.1 | 6 | 664 | |
NM_001102437.1 | NP_001095907.1 | 6 | 791 | ||
NM_001102438.1 | NP_001095908.1 | 6 | 791 | ||
NM_028793.3 | NP_083069.1 | 6 | 664 | ||
GenBank mRNA | AK005001.2 | - | 5 | 580 | |
AK050450.1 | - | 6 | 630 | ||
AK147839.1 | - | 6 | 791 | ||
BC035202.1 | - | 6 | 720 | ||
BC053518.1 | - | 6 | 758 | ||
BC061484.1 | - | 5 | 562 | 5355466 | |
BC084584.1 | - | 6 | 596 | 5257773 | |
BC090641.1 | - | 6 | 615 |