Gene Transcripts

Gene Symbol
Mrpl32
Entrez Gene ID
75398
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
0610033O15Rik
Chromosome Location
Chr. 13: 14610301 - 14613037 on Build GRCm38
UniGene ID
Mm.36511
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029271.2 NP_083547.1 3 420
GenBank mRNA AB049650.1 - 3 451
AF279255.1 - 3 397
AK002753.1 - 3 421
AK031791.1 - 1 2378
AK078324.1 - 3 438
BC037368.1 - 2 124
BC052214.1 - 3 455 30018135

Gene Symbol
Gm6578
Entrez Gene ID
625347
Gene Name
mitochondrial ribosomal protein L32 pseudogene
Gene Aliases
EG625347
Chromosome Location
Chr. 6: 12099518 - 12109580 on Build GRCm38
UniGene ID
Mm.389084
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC099426.1 - 4 630

View Full Product Details