Gene Transcripts

Gene Symbol
Arxes2
Entrez Gene ID
76976
Gene Name
adipocyte-related X-chromosome expressed sequence 2
Gene Aliases
2900062L11Rik, Spc22/23, Spcs3
Chromosome Location
Chr. X: 135993820 - 135995359 on Build GRCm38
UniGene ID
Mm.328305
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029823.2 NP_084099.1 1 281
GenBank mRNA AK013740.1 - 1 278
AK077432.1 - 1 255
BC030351.1 - 1 281
BY773138.1 - 1 282

Gene Symbol
Arxes1
Entrez Gene ID
76219
Gene Name
adipocyte-related X-chromosome expressed sequence 1
Gene Aliases
6530401D17Rik, Spc22/23, Spcs3
Chromosome Location
Chr. X: 136033367 - 136034946 on Build GRCm38
UniGene ID
Mm.482965
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029541.3 NP_083817.1 1 281
GenBank mRNA AK018307.1 - 1 281
BC016270.1 - 1 260

View Full Product Details