Gene Transcripts
- Gene Symbol
- Zdhhc20
- Entrez Gene ID
- 75965
- Gene Name
- zinc finger, DHHC domain containing 20
- Gene Aliases
- 5033406L14Rik, AI448102, B230110O18Rik, HK11
- Chromosome Location
- Chr. 14: 57832702 - 57890834 on Build GRCm38
- UniGene ID
- Mm.29044
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_029492.4 |
NP_083768.4
|
2, 3 |
235 |
|
GenBank mRNA |
AK017158.1 |
- |
2, 3 |
236 |
|
AK031184.1 |
- |
2, 3 |
194 |
|
AK049270.1 |
- |
2, 3 |
215 |
|
AK139216.1 |
- |
2, 3 |
194 |
|
AK140182.1 |
- |
2, 3 |
196 |
|
AK151840.1 |
- |
2, 3 |
250 |
|
AK163174.1 |
- |
2, 3 |
233 |
|
AK170017.1 |
- |
2, 3 |
253 |
|
AK170139.1 |
- |
2, 3 |
176 |
|
AY668951.1 |
- |
2, 3 |
153 |
|
BC019536.1 |
- |
2, 3 |
252 |
4039734 |
View Full Product Details