Gene Transcripts
- Gene Symbol
- TCOF1
- Entrez Gene ID
- 6949
- Gene Name
- treacle ribosome biogenesis factor 1
- Gene Aliases
- MFD1, TCS, TCS1, treacle
- Chromosome Location
- Chr. 5: 150357639 - 150400308 on Build GRCh38
- UniGene ID
- Hs.519672
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000356.3 |
NP_000347.2
|
4, 5 |
494 |
|
NM_001008657.2 |
NP_001008657.1
|
4, 5 |
494 |
|
NM_001135243.1 |
NP_001128715.1
|
4, 5 |
494 |
|
NM_001135244.1 |
NP_001128716.1
|
4, 5 |
494 |
|
NM_001135245.1 |
NP_001128717.1
|
4, 5 |
494 |
|
NM_001195141.1 |
NP_001182070.1
|
4, 5 |
494 |
|
XM_005268502.3 |
XP_005268559.1
|
4, 5 |
431 |
|
XM_005268503.3 |
XP_005268560.1
|
4, 5 |
431 |
|
XM_005268504.3 |
XP_005268561.1
|
4, 5 |
431 |
|
XM_005268505.3 |
XP_005268562.1
|
4, 5 |
431 |
|
XM_005268506.3 |
XP_005268563.1
|
4, 5 |
431 |
|
XM_005268507.3 |
XP_005268564.1
|
4, 5 |
431 |
|
XM_005268508.3 |
XP_005268565.1
|
4, 5 |
431 |
|
XM_005268509.3 |
XP_005268566.1
|
4, 5 |
431 |
|
XM_011537678.2 |
XP_011535980.1
|
4, 5 |
431 |
|
XM_017009792.1 |
XP_016865281.1
|
4, 5 |
431 |
|
XM_017009793.1 |
XP_016865282.1
|
4, 5 |
431 |
|
XM_017009794.1 |
XP_016865283.1
|
4, 5 |
431 |
|
XM_017009795.1 |
XP_016865284.1
|
4, 5 |
431 |
|
GenBank mRNA |
AK225284.1 |
- |
4, 5 |
419 |
|
AK303611.1 |
- |
4, 5 |
419 |
|
AK307586.1 |
- |
4, 5 |
419 |
|
AY460334.1 |
- |
4, 5 |
386 |
|
BC011764.2 |
- |
4, 5 |
415 |
3616898 |
BC014559.2 |
- |
4, 5 |
415 |
4025673 |
BC016144.1 |
- |
4, 5 |
414 |
|
BC027252.1 |
- |
4, 5 |
342 |
|
BC033093.1 |
- |
4, 5 |
415 |
4577077 |
DC377932.1 |
- |
4, 5 |
498 |
|
U40847.1 |
- |
4, 5 |
479 |
|
U76366.1 |
- |
4, 5 |
461 |
|
View Full Product Details