Gene Transcripts

Gene Symbol
Lbh
Entrez Gene ID
77889
Gene Name
limb-bud and heart
Gene Aliases
1810009F10Rik, 6720416L16Rik
Chromosome Location
Chr. 17: 72918305 - 72941946 on Build GRCm38
UniGene ID
Mm.154109
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029999.4 NP_084275.3 3 523
GenBank mRNA AF317517.1 - 3 506
AK020110.1 - 3 523
BC026827.1 - 3 505 4218560
BC052470.1 - 3 503 6813866

View Full Product Details