Gene Transcripts

Gene Symbol
Parp9
Entrez Gene ID
80285
Gene Name
poly (ADP-ribose) polymerase family, member 9
Gene Aliases
ARTD9, AW214463, BC003281, Bagl, Bal, PARP-9
Chromosome Location
Chr. 16: 35938470 - 35972621 on Build GRCm38
UniGene ID
Mm.49074
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030253.3 NP_084529.1 5 1741
GenBank mRNA AK037903.1 - 5 1308
AK050032.1 - 5 1185
BC003281.1 - 5 1183 3581841
BC010312.1 - 4 1166
BC070466.1 - 5 1741 30615225

View Full Product Details