Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_033037.3 | NP_149026.1 | 1, 2 | 378 | |
GenBank mRNA | AK004249.1 | - | 1, 2 | 392 | |
AK011032.1 | - | 1, 2 | 392 | ||
AK149582.1 | - | 1, 2 | 392 | ||
BC013638.1 | - | 1, 2 | 378 | 4194939 | |
BC020375.1 | - | 1, 2 | 378 |