Gene Transcripts

Gene Symbol
Gpank1
Entrez Gene ID
81845
Gene Name
G patch domain and ankyrin repeats 1
Gene Aliases
Bat-4, Bat4, D17H6S54E, G5
Chromosome Location
Chr. 17: 35121473 - 35124815 on Build GRCm38
UniGene ID
Mm.7405
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001128597.1 NP_001122069.1 2 686
NM_032460.2 NP_115849.2 1 484
GenBank mRNA AB221568.1 - 1 451
BC057902.1 - 2 645 5027029
BC120637.1 - 1 485 40129560
BC120639.1 - 1 485 40129562

View Full Product Details