Gene Transcripts

Gene Symbol
Sucnr1
Entrez Gene ID
84112
Gene Name
succinate receptor 1
Gene Aliases
Gpr91
Chromosome Location
Chr. 3: 60081869 - 60087566 on Build GRCm38
UniGene ID
Mm.125110
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_032400.2 NP_115776.2 2 863
GenBank mRNA AF295367.1 - 2 879
AK080866.1 - 2 877
BC096665.1 - 2 836 1365839

View Full Product Details