Gene Transcripts

Gene Symbol
Igsf9
Entrez Gene ID
93842
Gene Name
immunoglobulin superfamily, member 9
Gene Aliases
644ETD8, Dasm1, Kiaa1355-hp, NRT1, Ncaml, mKIAA1355
Chromosome Location
Chr. 1: 172481776 - 172498877 on Build GRCm38
UniGene ID
Mm.214530
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145800.1 NP_001139272.1 5 681
NM_033608.3 NP_291086.2 5 747
GenBank mRNA AF317839.1 - 5 747
AK053544.1 - 4 1298
AK078536.1 - 5 675
AK145121.1 - 5 656
BC034594.1 - 5 663
BC040281.1 - 5 707 5364073

View Full Product Details