Gene Transcripts

Gene Symbol
Trim8
Entrez Gene ID
93679
Gene Name
tripartite motif-containing 8
Gene Aliases
AA408830, BB006755, GERP, Rnf27
Chromosome Location
Chr. 19: 46501648 - 46516805 on Build GRCm38
UniGene ID
Mm.392177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053100.2 NP_444330.2 2 1407
GenBank mRNA AF220035.1 - 1 26
AF281047.1 - 2 756
AK052763.1 - 2 142
AK155303.1 - 2 1354
BC029283.1 - 2 1341
BC037065.1 - 2 692
BC060094.1 - 2 637
BC082298.1 - 2 1282
BC152343.1 - 2 743 40093266
BC152344.1 - 2 743 40093269

View Full Product Details