Gene Transcripts

Gene Symbol
Mtmr4
Entrez Gene ID
170749
Gene Name
myotubularin related protein 4
Gene Aliases
AA596759, ESTM44, FYVE-DSP2, ZFYVE11, mKIAA0647
Chromosome Location
Chr. 11: 87592135 - 87616302 on Build GRCm38
UniGene ID
Mm.399376
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133215.1 NP_573478.1 3, 4 165
GenBank mRNA AF262986.1 - 3, 4 165
AK146641.1 - 3, 4 182
BC058091.1 - 3, 4 163 6822365
BC058364.1 - 3, 4 163 6820754

View Full Product Details