Gene Transcripts

Gene Symbol
Slc5a4a
Entrez Gene ID
64452
Gene Name
solute carrier family 5, member 4a
Gene Aliases
SGLT3-a
Chromosome Location
Chr. 10: 76147451 - 76189265 on Build GRCm38
UniGene ID
Mm.154797
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133184.2 NP_573447.2 14 1789
GenBank mRNA AF251267.1 - 14 1789
AF498304.1 - 3 360

View Full Product Details