Gene Transcripts

Gene Symbol
Glcci1
Entrez Gene ID
170772
Gene Name
glucocorticoid induced transcript 1
Gene Aliases
2310047L21Rik, A130036A18Rik, Fam117c, Gig18, Tssn1
Chromosome Location
Chr. 6: 8509600 - 8597549 on Build GRCm38
UniGene ID
Mm.210787
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286728.1 NP_001273657.1 8 1645
NM_001286729.1 NP_001273658.1 8 1642
NM_133236.3 NP_573499.2 8 2318
GenBank mRNA AK037672.1 - 1 404
AK088359.1 - 1 340
AY098636.1 - 8 2364
BC037750.1 - 1 828

View Full Product Details