Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000624.5 | NP_000615.3 | 3 | 380 | |
GenBank mRNA | AK096131.1 | - | 2, 3 | 316 | |
AK310869.1 | - | 2 | 255 | ||
AL080185.1 | - | 1 | 64 | ||
BC008915.2 | - | 3 | 275 | 3618167 | |
DC394575.1 | - | 2 | 240 | ||
HY003178.1 | - | 2 | 284 | ||
J02639.1 | - | 2 | 191 | ||
S58545.1 | - | 1 | 67 |