Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_133733.4 | NP_598494.2 | 2 | 460 | |
GenBank mRNA | AB040490.1 | - | 2 | 377 | |
AK028983.1 | - | 2 | 431 | ||
AK033723.1 | - | 2 | 462 | ||
AK080481.1 | - | 2 | 415 | ||
AY259213.1 | - | 2 | 89 | ||
AY326421.1 | - | 2 | 95 | ||
BC026447.1 | - | 2 | 286 | 4459717 |