Gene Transcripts

Gene Symbol
Ubac1
Entrez Gene ID
98766
Gene Name
ubiquitin associated domain containing 1
Gene Aliases
1110033G07Rik, AA407978, GBDR1, Gdbr1, Ubadc1
Chromosome Location
Chr. 2: 25996958 - 26021781 on Build GRCm38
UniGene ID
Mm.247824
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_133835.2 NP_598596.2 1 354
GenBank mRNA AK157857.1 - 1 188
BC021811.1 - 1 305 3662474
CX243095.1 - 1 354

View Full Product Details