Gene Transcripts

Gene Symbol
Vmn1r18
Entrez Gene ID
171199
Gene Name
vomeronasal 1 receptor 18
Gene Aliases
V1rc26
Chromosome Location
Chr. 6: 57389668 - 57390567 on Build GRCm38
UniGene ID
Mm.377177
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134181.1 NP_598942.1 1 543
GenBank mRNA BC132061.1 - 1 642
BC138409.1 - 1 642 8861429

View Full Product Details