Gene Transcripts
- Gene Symbol
- Usp39
- Entrez Gene ID
- 28035
- Gene Name
- ubiquitin specific peptidase 39
- Gene Aliases
- AA408960, AI894154, CGI-21, D6Wsu157e, SAD1
- Chromosome Location
- Chr. 6: 72318676 - 72345217 on Build GRCm38
- UniGene ID
- Mm.281900
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_138592.4 |
NP_613058.1
|
4, 5 |
592 |
|
GenBank mRNA |
AK010443.1 |
- |
4, 5 |
602 |
|
AK018595.1 |
- |
4, 5 |
593 |
|
AK033105.1 |
- |
4, 5 |
589 |
|
AK045062.1 |
- |
4, 5 |
589 |
|
AK088354.1 |
- |
4, 5 |
604 |
|
AK165625.1 |
- |
4, 5 |
573 |
|
AK167668.1 |
- |
4, 5 |
580 |
|
AK168003.1 |
- |
4, 5 |
599 |
|
BC026983.1 |
- |
4, 5 |
567 |
5365862 |
View Full Product Details