Gene Transcripts

Gene Symbol
SPP1
Entrez Gene ID
6696
Gene Name
secreted phosphoprotein 1
Gene Aliases
BNSP, BSPI, ETA-1, OPN
Chromosome Location
Chr. 4: 87975650 - 87983411 on Build GRCh38
UniGene ID
Hs.313
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000582.2 NP_000573.1 5 634
NM_001040058.1 NP_001035147.1 6 676
NM_001040060.1 NP_001035149.1 5 595
NM_001251829.1 NP_001238758.1 4 553
NM_001251830.1 NP_001238759.1 7 865
XM_017008564.1 XP_016864053.1 4 1102
GenBank mRNA AB209987.1 - 3 388
AB469789.1 - 3 388
AF052124.1 - 5 556
AK057738.1 - 6 612
AK075463.1 - 7 546
AK290090.1 - 5 531
AK290104.1 - 6 613
AK295491.1 - 7 800
AK296035.1 - 5 531
AK315461.1 - 5 564
BC007016.1 - 5 570 4052438
BC017387.1 - 6 616 3828885
BC022844.1 - 5 571 4284921
BC093033.1 - 5 587 5288027
BX648003.1 - 5 549
D28759.1 - 5 511
D28760.1 - 4 469
D28761.1 - 4 430
DQ839491.1 - 5 511
DQ846870.1 - 5 511
DQ846871.1 - 5 511
J04765.1 - 5 577
JF412666.1 - 6 447
JF412667.1 - 5 593
KR062183.1 - 7 800
M83248.1 - 6 608
X13694.1 - 6 578

View Full Product Details