Gene Transcripts

Gene Symbol
Slc6a20a
Entrez Gene ID
102680
Gene Name
solute carrier family 6 (neurotransmitter transporter), member 20A
Gene Aliases
A730081N20Rik, AU022428, Xt3s1, Xtrp3s1
Chromosome Location
Chr. 9: 123636907 - 123678832 on Build GRCm38
UniGene ID
Mm.440867
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AJ428067.1 - 2332
AK080979.1 - 2332
AK140445.1 - 2330
AK160468.1 - 2330

View Full Product Details