Gene Transcripts

Gene Symbol
Kazn
Entrez Gene ID
71529
Gene Name
kazrin, periplakin interacting protein
Gene Aliases
2310007B04Rik, 9030409G11Rik, AI256607, AI666764, AW061099, Kaz, Kazrin
Chromosome Location
Chr. 4: 142102390 - 142239401 on Build GRCm38
UniGene ID
Mm.255986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_144531.3 NP_653114.3 14 3453
GenBank mRNA AK032460.1 - 12 2692
AK173090.1 - 14 3453

View Full Product Details