Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145209.3 | NP_660210.1 | 2 | 237 | |
GenBank mRNA | AB067533.1 | - | 1 | 162 | |
AF426289.1 | - | 1 | 168 | ||
AK078690.1 | - | 2 | 238 | ||
AK089111.1 | - | 1 | 207 | ||
AK149824.1 | - | 1 | 161 | ||
AK150863.1 | - | 1 | 291 | ||
AK165578.1 | - | 1 | 274 | ||
AY057107.1 | - | 1 | 150 | ||
AY089728.1 | - | 1 | 162 | ||
BC032152.1 | - | 1 | 174 | 5068699 |