Gene Transcripts
- Gene Symbol
- Appl1
- Entrez Gene ID
- 72993
- Gene Name
- adaptor protein, phosphotyrosine interaction, PH domain and leucine zipper containing 1
- Gene Aliases
- 2900057D21Rik, 7330406P05Rik, AI585782, AW209077, BB022931, C88264, DIP13
- Chromosome Location
- Chr. 14: 26918986 - 26971205 on Build GRCm38
- UniGene ID
- Mm.202322
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_145221.2 |
NP_660256.1
|
4, 5 |
378 |
|
GenBank mRNA |
AK013715.1 |
- |
4, 5 |
375 |
|
AK030445.1 |
- |
4, 5 |
258 |
|
AK033566.1 |
- |
4, 5 |
349 |
|
AK045438.1 |
- |
4, 5 |
314 |
|
AK049307.1 |
- |
4, 5 |
365 |
|
AK088080.1 |
- |
4, 5 |
329 |
|
AK146356.1 |
- |
4, 5 |
349 |
|
AY113705.1 |
- |
4, 5 |
343 |
|
BC063751.1 |
- |
4, 5 |
390 |
6511401 |
BU514763.1 |
- |
4, 5 |
389 |
|
View Full Product Details