Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001110512.1 | NP_001103982.1 | 9 | 912 | |
NM_145524.3 | NP_663499.2 | 10 | 1376 | ||
GenBank mRNA | AK087927.1 | - | 9 | 1256 | |
BC004636.1 | - | 11 | 1423 | 3585719 | |
BC057960.1 | - | 10 | 1243 | 5354784 | |
DQ093357.1 | - | 8 | 1237 | ||
DQ093358.1 | - | 10 | 1352 | ||
DQ230973.1 | - | 9 | 1144 | ||
DQ230974.1 | - | 9 | 916 |