Gene Transcripts

Gene Symbol
Dusp15
Entrez Gene ID
252864
Gene Name
dual specificity phosphatase-like 15
Gene Aliases
AI851682, LMW-DSP10, T-DSP10
Chromosome Location
Chr. 2: 152940995 - 152952277 on Build GRCm38
UniGene ID
Mm.330671
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001159376.1 NP_001152848.1 3, 4 373
NM_145744.2 NP_665687.1 3, 4 196
GenBank mRNA AF357887.1 - 3, 4 196
BC116841.1 - 3, 4 191 40126160
BC116843.1 - 3, 4 191 40126162
BC144711.1 - 2, 3 157

View Full Product Details