Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_146126.4 | NP_666238.1 | 7 | 704 | |
GenBank mRNA | AK004692.1 | - | 7 | 728 | |
AK015059.1 | - | 7 | 705 | ||
AK152169.1 | - | 7 | 753 | ||
AK166988.1 | - | 7 | 704 | ||
AK166996.1 | - | 7 | 733 | ||
BC024124.1 | - | 7 | 719 | 5099395 | |
BC030875.1 | - | 7 | 693 | 4237164 | |
BC039957.1 | - | 7 | 704 | ||
BC092291.1 | - | 7 | 707 | 5097505 | |
U27014.1 | - | 704 |