Gene Transcripts

Gene Symbol
SLC2A4
Entrez Gene ID
6517
Gene Name
solute carrier family 2 member 4
Gene Aliases
GLUT4
Chromosome Location
Chr. 17: 7281735 - 7288048 on Build GRCh38
UniGene ID
Hs.380691
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042.2 NP_001033.1 2, 3 358
GenBank mRNA AK300899.1 - 2, 3 425
AK313372.1 - 2, 3 372
BC034387.1 - 2, 3 358 5187454
BC069615.1 - 2, 3 253 7262457
BC069621.1 - 2, 3 253 7262469
BC113592.1 - 2, 3 185 8322644
BC126164.1 - 2, 3 185 8991880
M20747.1 - 2, 3 303

View Full Product Details