Gene Transcripts

Gene Symbol
RPL38
Entrez Gene ID
6169
Gene Name
ribosomal protein L38
Gene Aliases
L38
Chromosome Location
Chr. 17: 74203656 - 74209880 on Build GRCh38
UniGene ID
Hs.380953
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000999.3 NP_000990.1 4 292
NM_001035258.1 NP_001030335.1 4 257
GenBank mRNA AK312119.1 - 4 260
BC000603.2 - 4 229 3346447
BQ276548.1 - 4 288
BU532782.1 - 4 260
CB124619.1 - 4 282
CB128529.1 - 4 235
Z26876.1 - 4 274

View Full Product Details