Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010.2 | NP_001001.2 | 3 | 310 | |
GenBank mRNA | AB062123.1 | - | 3 | 296 | |
AK093634.1 | - | 1511 | |||
AK291517.1 | - | 3 | 309 | ||
AK311861.1 | - | 3 | 307 | ||
BC000524.2 | - | 2 | 272 | 2961137 | |
BC009427.2 | - | 2 | 263 | ||
BC013296.2 | - | 3 | 293 | 3030319 | |
BC027620.1 | - | 3 | 313 | 4698843 | |
BC035447.1 | - | 1 | 134 | ||
BC071907.1 | - | 3 | 298 | 4900361 | |
BC071908.1 | - | 3 | 298 | 5089456 | |
BC094826.1 | - | 3 | 294 | 5481378 | |
J03537.1 | - | 2 | 268 | ||
M20020.1 | - | 3 | 295 |