Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153541.3 | NP_705769.2 | 2 | 201 | |
GenBank mRNA | AK132152.1 | - | 2 | 239 | |
AK140914.1 | - | 2 | 190 | ||
AK163930.1 | - | 2 | 202 | ||
AK163989.1 | - | 2 | 205 | ||
BC023839.1 | - | 2 | 196 | 5344921 |