Gene Transcripts

Gene Symbol
Ammecr1l
Entrez Gene ID
225339
Gene Name
AMME chromosomal region gene 1-like
Gene Aliases
5430429D03Rik, AU022236, AU040755, AW111353, E230022H04Rik
Chromosome Location
Chr. 18: 31759475 - 31784083 on Build GRCm38
UniGene ID
Mm.290804
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242430.1 NP_001229359.1 4 1222
NM_153515.4 NP_705735.1 4 1021
GenBank mRNA AK030009.1 - 4 1229
AK054131.1 - 4 1020
AK133018.1 - 4 1223
AK151060.1 - 4 1029
AK151257.1 - 4 929
BC012310.1 - 3 626
BC034661.1 - 4 762 5137474
BC056948.1 - 4 965 6827318

View Full Product Details