Gene Transcripts
- Gene Symbol
- Wdr37
- Entrez Gene ID
- 207615
- Gene Name
- WD repeat domain 37
- Gene Aliases
- Chromosome Location
- Chr. 13: 8802966 - 8871904 on Build GRCm38
- UniGene ID
- Mm.284654
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039388.2 |
NP_001034477.1
|
2, 3 |
435 |
|
NM_001039389.2 |
NP_001034478.1
|
2, 3 |
356 |
|
NM_172445.3 |
NP_766033.1
|
2, 3 |
356 |
|
GenBank mRNA |
AK014128.1 |
- |
2, 3 |
332 |
|
AK032582.1 |
- |
2, 3 |
318 |
|
AK036203.1 |
- |
2, 3 |
352 |
|
AK044740.1 |
- |
2, 3 |
336 |
|
AK129254.1 |
- |
2, 3 |
312 |
|
AK146209.1 |
- |
2, 3 |
331 |
|
BB611667.1 |
- |
2, 3 |
435 |
|
BC046236.1 |
- |
1, 2 |
183 |
6817089 |
BC048819.1 |
- |
2, 3 |
280 |
|
CJ134699.1 |
- |
2, 3 |
357 |
|
View Full Product Details