Gene Transcripts

Gene Symbol
Zdhhc17
Entrez Gene ID
320150
Gene Name
zinc finger, DHHC domain containing 17
Gene Aliases
A230053P19Rik, BB187739, D130071N24Rik, Hip14
Chromosome Location
Chr. 10: 110941779 - 111010066 on Build GRCm38
UniGene ID
Mm.339281
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172554.2 NP_766142.2 4 381
GenBank mRNA AK038670.1 - 4 325
AK038681.1 - 4 325
AK039575.1 - 4 382
AK046891.1 - 4 323
AK122406.1 - 4 455
AK134967.1 - 4 330
AK162922.1 - 4 319
BC027101.1 - 4 310
BC058772.1 - 4 322
BC080664.1 - 4 320
BC117788.1 - 4 323 40092137
BC117789.2 - 4 323 40092140

View Full Product Details