Gene Transcripts

Gene Symbol
GAS8-AS1
Entrez Gene ID
750
Gene Name
GAS8 antisense RNA 1
Gene Aliases
C16orf3
Chromosome Location
Chr. 16: 90028908 - 90029901 on Build GRCh38
UniGene ID
Hs.633032
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_122031.1 1 796
GenBank mRNA AF050080.1 - 1 237
AF050081.1 - 1 819
BC093751.1 - 1 262 7939596
BC104849.1 - 1 262 8143852

View Full Product Details